missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluorobenzaldehyde, 98%

$131.76 - $131.76

Chemical Identifiers

CAS 459-57-4
Molecular Formula C7H5FO
Formula Weight 124.11
MDL Number MFCD00003378
UN Number UN1989
Beilstein 385857
Chemical Name or Material 4-Fluorobenzaldehyde
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1538314 SDS Alfa Aesar™
A1538314
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAA1538322 SDS Alfa Aesar™
A1538322
100g Each for $131.76
AAA1538336 SDS Alfa Aesar™
A1538336
500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
4-Fluorobenzaldehyde is used in the preparation of pyrazolopyridine derivative, which finds application as mitogen-activated protein kinase (MAPK) inhibitor. It also serves as a synthetic intermediate in the preparation of pharmaceutical compounds.

Solubility
Immiscible with water.

Notes
Air sensitive. Incompatible with strong bases, strong oxidizing agents and strong reducing agents.
Description & Specifications

Chemical Identifiers

459-57-4
124.11
UN1989
4-Fluorobenzaldehyde
C7H5FO
MFCD00003378
385857

Specifications

56°C (133°F)
−10°C
459-57-4
124.11
UN1989
4-Fluorobenzaldehyde
180° to 181°C
25g
C7H5FO
MFCD00003378
385857
98%

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, N.O.S.

TSCA : TSCA