missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoroanisole, 99%

Chemical Identifiers

CAS 459-60-9
Molecular Formula C7H7FO
Formula Weight 126.13
MDL Number MFCD00000348
UN Number UN1993
Beilstein 1281425
Synonym 4-Methoxyfluorobenzene
Chemical Name or Material 4-Fluoroanisole
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2304814 SDS Alfa Aesar™
B2304814
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAB2304822 SDS Alfa Aesar™
B2304822
100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
4-Fluoroanisole is a raw material for medicine and intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
Description & Specifications

Chemical Identifiers

459-60-9
126.13
UN1993
4-Methoxyfluorobenzene
C7H7FO
MFCD00000348
1281425
4-Fluoroanisole

Specifications

43°C (109°F)
−45°C
459-60-9
126.13
UN1993
4-Methoxyfluorobenzene
99%
156° to 158°C
25g
C7H7FO
MFCD00000348
1281425
4-Fluoroanisole

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA