Learn More
4-Fluoroanisole, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2304822
Description
4-Fluoroanisole is a raw material for medicine and intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Fluoroanisole is a raw material for medicine and intermediate.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
Specifications
4-Fluoroanisole | |
-45°C | |
156°C to 158°C | |
C7H7FO | |
MFCD00000348 | |
UN1993 | |
4-fluoroanisole, p-fluoroanisole, benzene, 1-fluoro-4-methoxy, p-fluoromethoxybenzene, p-methoxyfluorobenzene, 4-fluoroanisol, p-fluorophenyl methyl ether, anisole, p-fluoro, 1-fluoro-4-methoxy-benzene, 4-methoxyfluorobenzene | |
VIPWUFMFHBIKQI-UHFFFAOYSA-N | |
1-fluoro-4-methoxybenzene | |
9987 | |
99% |
459-60-9 | |
1.116 | |
43°C (109°F) | |
1.488 | |
100 g | |
1281425 | |
Not miscible or difficult to mix in water. | |
COC1=CC=C(C=C1)F | |
126.13 | |
126.13 |
Chemical Identifiers
459-60-9 | |
126.13 | |
VIPWUFMFHBIKQI-UHFFFAOYSA-N | |
9987 | |
COC1=CC=C(C=C1)F |
C7H7FO | |
MFCD00000348 | |
4-fluoroanisole, p-fluoroanisole, benzene, 1-fluoro-4-methoxy, p-fluoromethoxybenzene, p-methoxyfluorobenzene, 4-fluoroanisol, p-fluorophenyl methyl ether, anisole, p-fluoro, 1-fluoro-4-methoxy-benzene, 4-methoxyfluorobenzene | |
1-fluoro-4-methoxybenzene |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 207-295-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only