Learn More
4'-Fluoroacetophenone, 99%, ACROS Organics™
Manufacturer: Fisher Scientific Acros 119261000
Specifications
4'-Fluoroacetophenone | |
71°C | |
Glass bottle | |
1.13 | |
Colorless to Yellow | |
100mL | |
403-42-9 | |
100.0 | |
C8H7FO | |
MFCD00000354 | |
ZDPAWHACYDRYIW-UHFFFAOYSA-N | |
1-(4-fluorophenyl)ethanone | |
9828 | |
Liquid |
1.1300g/mL | |
Authentic | |
1.5090 to 1.512 | |
196°C | |
4°C | |
99% | |
98.5 | |
98.5% min. (GC) | |
FC6H4COCH3 | |
1-4-fluorophenyl ethan-1-one, 1-4-fluorophenyl ethanone, 1-acetyl-4-fluorobenzene, 4 fluoroacetophenone, 4'-fluoroacetophenone, 4-fluoroacetophenone, acetophenone, 4'-fluoro, ethanone, 1-4-fluorophenyl, p-fluoroacetophenone, unii-tvq090602v | |
CC(=O)C1=CC=C(C=C1)F | |
138.14 | |
138.14 | |
99% |
Chemical Identifiers
403-42-9 | |
138.14 | |
ZDPAWHACYDRYIW-UHFFFAOYSA-N | |
9828 | |
CC(=O)C1=CC=C(C=C1)F |
C8H7FO | |
MFCD00000354 | |
1-4-fluorophenyl ethan-1-one, 1-4-fluorophenyl ethanone, 1-acetyl-4-fluorobenzene, 4 fluoroacetophenone, 4'-fluoroacetophenone, 4-fluoroacetophenone, acetophenone, 4'-fluoro, ethanone, 1-4-fluorophenyl, p-fluoroacetophenone, unii-tvq090602v | |
1-(4-fluorophenyl)ethanone |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 206-960-9
TSCA : TSCA