Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Fluoroacetanilide, 98%

$38.18 - $129.92

Chemical Identifiers

Chemical Name or Material 4'-Fluoroacetanilide
CAS 351-83-7
Molecular Formula C8H8FNO
Formula Weight 153.16
MDL Number MFCD00016335
Beilstein 2208090
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1897414

 
alfa aesar™
A1897414
25g Each for $38.18

AAA1897422

 
alfa aesar™
A1897422
100g Each for $129.92
Description & Specifications

Specifications

Formula Weight 153.16
Chemical Name or Material 4'-Fluoroacetanilide
Assay 98%
Molecular Formula C8H8FNO
MDL Number MFCD00016335
CAS 351-83-7
Beilstein 2208090
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number AE2977000
Melting Point 151° to 153°C
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.