missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluoro-o-cresol, TCI America™

Manufacturer:  TCI America F038725G

 View more versions of this product

Catalog No. F038725G



Specifications

Specifications

4-Fluoro-o-cresol
Yellow
25g
C7H7FO
2-methyl-4-fluorophenol, 4-fluoro-o-cresol, 4-fluoro-2-methyl-phenol, 5-fluoro-2-hydroxytoluene, phenol, 4-fluoro-2-methyl, pubchem1500, acmc-1csjj, 4-fuoro-2-methyl-phenol, 2-methyl-4-fluoro phenol, 4-fluoro-2-methyl phenol
CC1=C(C=CC(=C1)F)O
126.13
126.13
≥98.0% (GC)
93°C
36°C
452-72-2
MFCD00075088
GKQDDKKGDIVDAG-UHFFFAOYSA-N
4-fluoro-2-methylphenol
136295
Crystal at 20°C

Chemical Identifiers

452-72-2
126.13
GKQDDKKGDIVDAG-UHFFFAOYSA-N
136295
CC1=C(C=CC(=C1)F)O
C7H7FO
MFCD00075088
2-methyl-4-fluorophenol, 4-fluoro-o-cresol, 4-fluoro-2-methyl-phenol, 5-fluoro-2-hydroxytoluene, phenol, 4-fluoro-2-methyl, pubchem1500, acmc-1csjj, 4-fuoro-2-methyl-phenol, 2-methyl-4-fluoro phenol, 4-fluoro-2-methyl phenol
4-fluoro-2-methylphenol
Safety and Handling

Safety and Handling

TSCA : No

Documents