missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-N,N-dimethyl-2-nitroaniline, 97%

Manufacturer:  Alfa Aesar™ H6335706

 View more versions of this product

Catalog No. AAH6335706



Specifications

Specifications

4-Fluoro-N,N-dimethyl-2-nitroaniline
69261-01-4
MFCD04116334
n,n-dimethyl 4-fluoro-2-nitroaniline, n,n-dimethyl-4-fluoro-2-nitroaniline, benzenamine,4-fluoro-n,n-dimethyl-2-nitro, pubchem3366, acmc-1b82t, 4-fluoro-2-nitrophenyl dimethylamine, 4-fluoranyl-n,n-dimethyl-2-nitro-aniline
CN(C)C1=C(C=C(C=C1)F)[N+](=O)[O-]
184.17
184.17
97%
5g
C8H9FN2O2
UN2811
RGCUVPLVTHHXMT-UHFFFAOYSA-N
4-fluoro-N,N-dimethyl-2-nitroaniline
16440018
Solid

Chemical Identifiers

69261-01-4
184.17
RGCUVPLVTHHXMT-UHFFFAOYSA-N
16440018
CN(C)C1=C(C=C(C=C1)F)[N+](=O)[O-]
C8H9FN2O2
MFCD04116334
n,n-dimethyl 4-fluoro-2-nitroaniline, n,n-dimethyl-4-fluoro-2-nitroaniline, benzenamine,4-fluoro-n,n-dimethyl-2-nitro, pubchem3366, acmc-1b82t, 4-fluoro-2-nitrophenyl dimethylamine, 4-fluoranyl-n,n-dimethyl-2-nitro-aniline
4-fluoro-N,N-dimethyl-2-nitroaniline
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

SDS Documents