missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-N-methylaniline, 97%

Manufacturer:  Alfa Aesar™ H5510514

 View more versions of this product

Catalog No. AAH5510514



Description

Description

Applications
4-Fluoro-N-methylaniline is used as a model compound to study the in vivo and in vitro biotransformation of secondary aromatic amines.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents , acids, acid chloride, acid anhydride and chloroformates.
Specifications

Specifications

4-Fluoro-N-methylaniline
76°C (169°F)
79°C (11mmHg)
459-59-6
MFCD00017942
4-fluoro-n-methyl aniline, 4-fluoro-n-methyl-aniline, 4-fluoro-n-methylbenzenamine, 4-fluorophenyl-methylamine, acmc-1aem5, benzenamine, 4-fluoro-n-methyl, benzenamine,4-fluoro-n-methyl, n-methyl-4-fluoroaniline, unii-88f75yis0f
CNC1=CC=C(C=C1)F
125.146
125.15
1.04
1.532
25g
C7H8FN
UN2810
VLWRKVBQUANIGI-UHFFFAOYSA-N
4-fluoro-N-methylaniline
68024
97%

Chemical Identifiers

459-59-6
125.146
VLWRKVBQUANIGI-UHFFFAOYSA-N
68024
CNC1=CC=C(C=C1)F
C7H8FN
MFCD00017942
4-fluoro-n-methyl aniline, 4-fluoro-n-methyl-aniline, 4-fluoro-n-methylbenzenamine, 4-fluorophenyl-methylamine, acmc-1aem5, benzenamine, 4-fluoro-n-methyl, benzenamine,4-fluoro-n-methyl, n-methyl-4-fluoroaniline, unii-88f75yis0f
4-fluoro-N-methylaniline
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H317-H335-H227-H315
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.

P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

EINECSNumber : 207-294-1

TSCA : No

Recommended Storage : Ambient temperatures

Documents