Learn More
4-Fluoro-N-methylaniline, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5510503
Description
4-Fluoro-N-methylaniline is used as a model compound to study the biotransformation of secondary aromatic amines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Fluoro-N-methylaniline is used as a model compound to study the biotransformation of secondary aromatic amines.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents , acids, acid chloride, acid anhydride and chloroformates.
Specifications
4-Fluoro-N-methylaniline | |
1.04 | |
76°C (169°F) | |
1.532 | |
1 g | |
benzenamine, 4-fluoro-n-methyl, unii-88f75yis0f, n-methyl-4-fluoroaniline, acmc-1aem5, 4-fluoro-n-methyl aniline, 4-fluoro-n-methyl-aniline, 4-fluoro-n-methylbenzenamine, 4-fluorophenyl-methylamine, benzenamine,4-fluoro-n-methyl | |
CNC1=CC=C(C=C1)F | |
125.146 | |
125.15 |
459-59-6 | |
79°C (11 mmHg) | |
C7H8FN | |
MFCD00017942 | |
UN2810 | |
VLWRKVBQUANIGI-UHFFFAOYSA-N | |
4-fluoro-N-methylaniline | |
68024 | |
97% |
Chemical Identifiers
459-59-6 | |
125.146 | |
VLWRKVBQUANIGI-UHFFFAOYSA-N | |
4-fluoro-N-methylaniline |
C7H8FN | |
MFCD00017942 | |
68024 | |
CNC1=CC=C(C=C1)F |
Safety and Handling
GHS H Statement
H318-H302-H317-H335-H227-H315
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 207-294-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only