missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-N-(4-methoxybenzyl)aniline, 97%

$81.01 - $258.76

Chemical Identifiers

CAS 80143-71-1
Molecular Formula C14H14FNO
Molecular Weight (g/mol) 231.27
MDL Number MFCD00017946
InChI Key GSIXDRVRPADHPJ-UHFFFAOYSA-N
Synonym 4-fluoro-n-4-methoxybenzyl aniline
PubChem CID 4590297
IUPAC Name 4-fluoro-N-[(4-methoxyphenyl)methyl]aniline
SMILES COC1=CC=C(C=C1)CNC2=CC=C(C=C2)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH55731MD View Documents Alfa Aesar™
H55731MD
250mg Each for $81.01
 
AAH5573103 View Documents Alfa Aesar™
H5573103
1g Each for $258.76
 
Specifications

Chemical Identifiers

80143-71-1
231.27
GSIXDRVRPADHPJ-UHFFFAOYSA-N
4590297
COC1=CC=C(C=C1)CNC2=CC=C(C=C2)F
C14H14FNO
MFCD00017946
4-fluoro-n-4-methoxybenzyl aniline
4-fluoro-N-[(4-methoxyphenyl)methyl]aniline

Specifications

250mg
C14H14FNO
4-fluoro-n-4-methoxybenzyl aniline
COC1=CC=C(C=C1)CNC2=CC=C(C=C2)F
231.27
231.27
4-Fluoro-N-(4-methoxybenzyl)aniline
80143-71-1
MFCD00017946
GSIXDRVRPADHPJ-UHFFFAOYSA-N
4-fluoro-N-[(4-methoxyphenyl)methyl]aniline
4590297
97%
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures