missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-N-(4-fluorobenzyl)aniline, 97%

Manufacturer:  Alfa Aesar™ H56208MD

 View more versions of this product

Catalog No. AAH56208MD



Description

Description

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications

Specifications

4-Fluoro-N-(4-fluorobenzyl)aniline
80143-73-3
MFCD03210766
OQVIJNAAQBTAAY-UHFFFAOYSA-N
4-fluoro-N-[(4-fluorophenyl)methyl]aniline
1133337
97%
250mg
C13H11F2N
4-fluoro-n-4-fluorophenyl methyl aniline
C1=CC(=CC=C1CNC2=CC=C(C=C2)F)F
219.235
219.23

Chemical Identifiers

80143-73-3
219.235
OQVIJNAAQBTAAY-UHFFFAOYSA-N
1133337
C1=CC(=CC=C1CNC2=CC=C(C=C2)F)F
C13H11F2N
MFCD03210766
4-fluoro-n-4-fluorophenyl methyl aniline
4-fluoro-N-[(4-fluorophenyl)methyl]aniline
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents