Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-L-phenylglycine, 98%

Chemical Identifiers

Chemical Name or Material 4-Fluoro-L-phenylglycine
CAS 19883-57-9
Formula Weight 169.15
MDL Number MFCD00213799
Beilstein 5850631
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6149514

  SDS
alfa aesar™
H6149514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6149522

  SDS
alfa aesar™
H6149522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 169.15
MDL Number MFCD00213799
Assay 98%
Melting Point >300°C
Physical Form Solid
Chemical Name or Material 4-Fluoro-L-phenylglycine
Color White to off-white
Beilstein 5850631
CAS 19883-57-9
Quantity 25g

Applications
4-Fluoro-L-phenylglycine is an optically active amino acid derivative that is used as an intermediate for making drugs.

Solubility
Soluble in 1M HCl.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.