Please login to your online account to display your discounted pricing

4-Fluoro-DL-α-phenylglycine, 98+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Fluoro-DL-α-phenylglycine
CAS 7292-73-1
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C8H8FNO2
Linear Formula FC6H4CH(NH2)COOH
Formula Weight 169.15
MDL Number MFCD00066444
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC367930010

  SDS
fisher scientific acros
AC367930010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC367930050

  SDS
thermo fisher scientific
AC367930050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.0%
Formula Weight 169.15
Physical Form Powder
Quantity 1g
Color Pale yellow
CAS Max % 100.0
CAS Min % 98.0
Linear Formula FC6H4CH(NH2)COOH
Infrared Spectrum Authentic
Molecular Formula C8H8FNO2
Chemical Name or Material 4-Fluoro-DL-α-phenylglycine
MDL Number MFCD00066444
Packaging Glass bottle
Assay Percent Range 98+%
CAS 7292-73-1