missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Fluoro-DL-α-phenylglycine, 98+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 367930050
Specifications
4-Fluoro-DL-α-phenylglycine | |
98.0 | |
Authentic | |
Glass bottle | |
FC6H4CH(NH2)COOH | |
5g | |
JKFYKCYQEWQPTM-UHFFFAOYNA-N | |
2-amino-2-(4-fluorophenyl)acetic acid | |
98313 | |
98+% |
7292-73-1 | |
100.0 | |
98% min. (HPLC) | |
C8H8FNO2 | |
MFCD00066444,MFCD00042727 | |
2-amino-2-4-fluorophenyl acetic acid, 4-fluoro-dl-phenylglycine, 4-fluoro-dl-alpha-phenylglycine, dl-4-fluorophenylglycine, amino 4-fluorophenyl acetic acid, 4-fluorophenylglycine, dl-4-fluorophenyl glycine, amino-4-fluoro-phenyl-acetic acid, dl-alpha-4-fluorophenyl-glycine, benzeneacetic acid, alpha-amino-4-fluoro | |
NC(C(O)=O)C1=CC=C(F)C=C1 | |
169.16 | |
169.15 |
Chemical Identifiers
7292-73-1 | |
169.16 | |
JKFYKCYQEWQPTM-UHFFFAOYNA-N | |
98313 | |
NC(C(O)=O)C1=CC=C(F)C=C1 |
C8H8FNO2 | |
MFCD00066444,MFCD00042727 | |
2-amino-2-4-fluorophenyl acetic acid, 4-fluoro-dl-phenylglycine, 4-fluoro-dl-alpha-phenylglycine, dl-4-fluorophenylglycine, amino 4-fluorophenyl acetic acid, 4-fluorophenylglycine, dl-4-fluorophenyl glycine, amino-4-fluoro-phenyl-acetic acid, dl-alpha-4-fluorophenyl-glycine, benzeneacetic acid, alpha-amino-4-fluoro | |
2-amino-2-(4-fluorophenyl)acetic acid |
Safety and Handling
EINECSNumber : 230-724-4
RUO â Research Use Only