missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-DL-phenylglycine, 97%

$44.41 - $139.70

Chemical Identifiers

CAS 7292-73-1
Molecular Formula C8H8FNO2
Molecular Weight (g/mol) 169.155
MDL Number MFCD00066444
InChI Key JKFYKCYQEWQPTM-UHFFFAOYSA-N
Synonym 2-amino-2-4-fluorophenyl acetic acid, 4-fluoro-dl-phenylglycine, 4-fluoro-dl-alpha-phenylglycine, dl-4-fluorophenylglycine, amino 4-fluorophenyl acetic acid, 4-fluorophenylglycine, dl-4-fluorophenyl glycine, amino-4-fluoro-phenyl-acetic acid, dl-alpha-4-fluorophenyl-glycine, benzeneacetic acid, alpha-amino-4-fluoro
PubChem CID 98313
IUPAC Name 2-amino-2-(4-fluorophenyl)acetic acid
SMILES C1=CC(=CC=C1C(C(=O)O)N)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1893203 View Documents Alfa Aesar™
L1893203
1g Each for $44.41
 
AAL1893206 View Documents Alfa Aesar™
L1893206
5g Each for $139.70
 
Specifications

Chemical Identifiers

7292-73-1
169.155
JKFYKCYQEWQPTM-UHFFFAOYSA-N
98313
C1=CC(=CC=C1C(C(=O)O)N)F
C8H8FNO2
MFCD00066444
2-amino-2-4-fluorophenyl acetic acid, 4-fluoro-dl-phenylglycine, 4-fluoro-dl-alpha-phenylglycine, dl-4-fluorophenylglycine, amino 4-fluorophenyl acetic acid, 4-fluorophenylglycine, dl-4-fluorophenyl glycine, amino-4-fluoro-phenyl-acetic acid, dl-alpha-4-fluorophenyl-glycine, benzeneacetic acid, alpha-amino-4-fluoro
2-amino-2-(4-fluorophenyl)acetic acid

Specifications

7292-73-1
MFCD00066444
2-amino-2-4-fluorophenyl acetic acid, 4-fluoro-dl-phenylglycine, 4-fluoro-dl-alpha-phenylglycine, dl-4-fluorophenylglycine, amino 4-fluorophenyl acetic acid, 4-fluorophenylglycine, dl-4-fluorophenyl glycine, amino-4-fluoro-phenyl-acetic acid, dl-alpha-4-fluorophenyl-glycine, benzeneacetic acid, alpha-amino-4-fluoro
C1=CC(=CC=C1C(C(=O)O)N)F
169.155
169.16
>300°C
4-Fluoro-DL-phenylglycine
C8H8FNO2
2804849
JKFYKCYQEWQPTM-UHFFFAOYSA-N
2-amino-2-(4-fluorophenyl)acetic acid
98313
97%
1g