Alfa Aesar™ 4-Fluoro-DL-phenylglycine, 97%
Manufacturer:  Alfa Aesar™ L1893203
 View more versions of this product
Catalog No. AAL1893203
Specifications
Specifications
4-Fluoro-DL-phenylglycine
C8H8FNO2
2804849
JKFYKCYQEWQPTM-UHFFFAOYSA-N
2-amino-2-(4-fluorophenyl)acetic acid
98313
97%
1g
7292-73-1
MFCD00066444
2-amino-2-4-fluorophenyl acetic acid
C1=CC(=CC=C1C(C(=O)O)N)F
169.155
169.16
>300°C
Chemical Identifiers
7292-73-1
169.155
JKFYKCYQEWQPTM-UHFFFAOYSA-N
98313
C1=CC(=CC=C1C(C(=O)O)N)F
C8H8FNO2
MFCD00066444
2-amino-2-4-fluorophenyl acetic acid
2-amino-2-(4-fluorophenyl)acetic acid
Documents