Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-DL-glutamic acid, erythro + threo, 97%

$61.03 - $168.76

Chemical Identifiers

Chemical Name or Material 4-Fluoro-DL-glutamic acid
CAS 2708-77-2
Name Note erythro + threo
Molecular Formula C5H8FNO4
Formula Weight 165.13
MDL Number MFCD00055778
Beilstein 1947477
Synonym H-4-F-DL-Glu-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL10517MB

  SDS
alfa aesar™
L10517MB
25mg Each for $61.03

AAL10517MC

  SDS
alfa aesar™
L10517MC
100mg Each for $168.76
Description & Specifications

Specifications

Formula Weight 165.13
Chemical Name or Material 4-Fluoro-DL-glutamic acid
CAS 2708-77-2
MDL Number MFCD00055778
Synonym H-4-F-DL-Glu-OH
Assay 97%
Molecular Formula C5H8FNO4
Beilstein 1947477
Name Note erythro + threo
Melting Point ≈185°C
Quantity 25mg