missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-DL-glutamic acid, erythro + threo, 97%

Manufacturer:  Alfa Aesar™ L10517MC

 View more versions of this product

Catalog No. AAL10517MC



Specifications

Specifications

4-Fluoro-DL-glutamic acid
2708-77-2
MFCD00055778
4-fluoro-dl-glutamic acid, 4-fluoroglutamic acid, dl-erythro-4-fluoroglutamic acid, 2-azanyl-4-fluoranyl-pentanedioic acid, d-glutamic acid,4-fluoro-, 4r, 4-fluoroglutamate, dl-4-fluoroglutamic acid, acmc-1bws0, acmc-20n5m3, dl-glutamic acid, 4-fluoro
C(C(C(=O)O)N)C(C(=O)O)F
165.12
165.13
~185°C
erythro + threo
C5H8FNO4
1947477
JPSHPWJJSVEEAX-UHFFFAOYSA-N
2-amino-4-fluoropentanedioic acid
102871
97%
100mg

Chemical Identifiers

2708-77-2
165.12
JPSHPWJJSVEEAX-UHFFFAOYSA-N
102871
C(C(C(=O)O)N)C(C(=O)O)F
C5H8FNO4
MFCD00055778
4-fluoro-dl-glutamic acid, 4-fluoroglutamic acid, dl-erythro-4-fluoroglutamic acid, 2-azanyl-4-fluoranyl-pentanedioic acid, d-glutamic acid,4-fluoro-, 4r, 4-fluoroglutamate, dl-4-fluoroglutamic acid, acmc-1bws0, acmc-20n5m3, dl-glutamic acid, 4-fluoro
2-amino-4-fluoropentanedioic acid
Documents