Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-beta-nitrostyrene, 98%

Chemical Identifiers

Chemical Name or Material 4-Fluoro-beta-nitrostyrene
CAS 706-08-1
Formula Weight 167.14
MDL Number MFCD00024824
UN Number UN2811
Beilstein 2208372
Synonym 1-Fluoro-4-(2-nitrovinyl)benzene; 1-(4-Fluorophenyl)-2-nitroethylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6461306

  SDS
alfa aesar™
H6461306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6461314

  SDS
alfa aesar™
H6461314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6461322

  SDS
alfa aesar™
H6461322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 167.14
Melting Point 99° to 102°C
Synonym 1-Fluoro-4-(2-nitrovinyl)benzene; 1-(4-Fluorophenyl)-2-nitroethylene
Assay 98%
Health Hazard 1 Danger
Chemical Name or Material 4-Fluoro-beta-nitrostyrene
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P310-P301+P312-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 706-08-1
Quantity 5g
UN Number UN2811
Color Pale yellow
Beilstein 2208372
Physical Form Crystalline Solid
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
MDL Number MFCD00024824
Health Hazard 2 GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
Storage Note 1 2° to 8°C
RTECS Number WL4800000

Applications
It is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store at 99-102°C. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P280-P264-P305+P351+P338-P310-P301+P312-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.

2° to 8°C