Please login to your online account to display your discounted pricing

Alfa Aesar™ (4-Fluoro-alpha-tolulenesulfonyl)acetic acid, 98+%

$131.99 - $394.77

Chemical Identifiers

Chemical Name or Material (4-Fluoro-alpha-tolulenesulfonyl)acetic acid
CAS 222639-41-0
Molecular Formula C9H9FO4S
Formula Weight 232.24
MDL Number MFCD06208212
Synonym 2-(4-Fluorobenzylsulfonyl)acetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27573MC

  SDS
alfa aesar™
H27573MC
100mg Each for $131.99

AAH27573ME

  SDS
alfa aesar™
H27573ME
500mg Each for $394.77
Description & Specifications

Specifications

Formula Weight 232.24
Molecular Formula C9H9FO4S
MDL Number MFCD06208212
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material (4-Fluoro-alpha-tolulenesulfonyl)acetic acid
Melting Point ≈145°C
Health Hazard 1 Warning
Synonym 2-(4-Fluorobenzylsulfonyl)acetic acid
Assay ≥98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 222639-41-0
Quantity 100mg
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.