missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (4-Fluoro-alpha-toluenesulfonyl)acetamidoxime

$198.57 - $885.08

Chemical Identifiers

CAS 175276-85-4
Molecular Formula C9H11FN2O3S
Formula Weight 246.26
MDL Number MFCD00052932
Synonym 2-(4-Fluorobenzylsulfonyl)acetamidoxime
Chemical Name or Material (4-Fluoro-alpha-toluenesulfonyl)acetamidoxime
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5065103 SDS Alfa Aesar™
H5065103
1g Each for $198.57
AAH5065106 SDS Alfa Aesar™
H5065106
5g Each for $885.08
Specifications Description & Specifications

Chemical Identifiers

175276-85-4
246.26
2-(4-Fluorobenzylsulfonyl)acetamidoxime
C9H11FN2O3S
MFCD00052932
(4-Fluoro-alpha-toluenesulfonyl)acetamidoxime

Specifications

1g
C9H11FN2O3S
MFCD00052932
(4-Fluoro-alpha-toluenesulfonyl)acetamidoxime
175276-85-4
246.26
2-(4-Fluorobenzylsulfonyl)acetamidoxime

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning