missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluoro-alpha-methylstyrene (stabilized with TBC), TCI America™

Manufacturer:  TCI America F02505ML

 View more versions of this product

Catalog No. F02505ML



Specifications

Specifications

4-Fluoro-alpha-methylstyrene (stabilized with TBC)
Undesignated
350-40-3
MFCD00042297
4-fluoro-alpha-methylstyrene, 1-fluoro-4-1-methylethenyl benzene, 1-fluoro-4-prop-1-en-2-yl benzene, 2-4-fluorophenyl-1-propene, 2-4-fluorophenyl propene, benzene, 1-fluoro-4-1-methylethenyl, 4-fluoro-.alpha.-methylstyrene, 1-fluoro-4-isopropenyl benzene, 1-fluoro-4-isopropenylbenzene, p-fluoro-.alpha.-methylstyrene
CC(=C)C1=CC=C(C=C1)F
136.169
136.17
≥98.0% (GC)
183°C
5mL
C9H9F
1993
VIXHMBLBLJSGIB-UHFFFAOYSA-N
1-fluoro-4-prop-1-en-2-ylbenzene
67690
Clear Liquid at 20°C

Chemical Identifiers

350-40-3
136.169
VIXHMBLBLJSGIB-UHFFFAOYSA-N
67690
CC(=C)C1=CC=C(C=C1)F
C9H9F
MFCD00042297
4-fluoro-alpha-methylstyrene, 1-fluoro-4-1-methylethenyl benzene, 1-fluoro-4-prop-1-en-2-yl benzene, 2-4-fluorophenyl-1-propene, 2-4-fluorophenyl propene, benzene, 1-fluoro-4-1-methylethenyl, 4-fluoro-.alpha.-methylstyrene, 1-fluoro-4-isopropenyl benzene, 1-fluoro-4-isopropenylbenzene, p-fluoro-.alpha.-methylstyrene
1-fluoro-4-prop-1-en-2-ylbenzene
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents