missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
4-Fluoro-5-nitrobenzene-1,2-diamine, Technical Grade, Thermo Scientific™
Chemical Identifiers
CAS |
113269-06-0 |
Molecular Formula |
C6H6FN3O2 |
Molecular Weight (g/mol) |
171.131 |
InChI Key |
QZNALIMEKAACKP-UHFFFAOYSA-N |
Synonym |
1,2-benzenediamine,4-fluoro-5-nitro, 1,2-benzenediamine, 4-fluoro-5-nitro, acmc-20ag30, 4,5-diamine-2-fluoronitrobenzene, 5-fluoro-4-nitrobenzene-1,2-diamine, 4-fluoro-5-nitro-1,2-phenylenediamine |
PubChem CID |
3782733 |
IUPAC Name |
4-fluoro-5-nitrobenzene-1,2-diamine |
SMILES |
C1=C(C(=CC(=C1[N+](=O)[O-])F)N)N |
View More Specs
Products
2
Description
Specifications
Chemical Identifiers
|
113269-06-0 |
|
171.131 |
|
1,2-benzenediamine,4-fluoro-5-nitro, 1,2-benzenediamine, 4-fluoro-5-nitro, acmc-20ag30, 4,5-diamine-2-fluoronitrobenzene, 5-fluoro-4-nitrobenzene-1,2-diamine, 4-fluoro-5-nitro-1,2-phenylenediamine |
|
4-fluoro-5-nitrobenzene-1,2-diamine |
|
C6H6FN3O2 |
|
QZNALIMEKAACKP-UHFFFAOYSA-N |
|
3782733 |
|
C1=C(C(=CC(=C1[N+](=O)[O-])F)N)N |
Specifications
|
113269-06-0 |
|
C6H6FN3O2 |
|
1,2-benzenediamine,4-fluoro-5-nitro, 1,2-benzenediamine, 4-fluoro-5-nitro, acmc-20ag30, 4,5-diamine-2-fluoronitrobenzene, 5-fluoro-4-nitrobenzene-1,2-diamine, 4-fluoro-5-nitro-1,2-phenylenediamine |
|
C1=C(C(=CC(=C1[N+](=O)[O-])F)N)N |
|
171.131 |
|
171.13 |
|
Technical |
|
Amber Glass Bottle |
|
1g |
|
QZNALIMEKAACKP-UHFFFAOYSA-N |
|
4-fluoro-5-nitrobenzene-1,2-diamine |
|
3782733 |
|
Tech. |
|
4-Fluoro-5-nitrobenzene-1, 2-diamine |
SDS
missing translation for 'documents'