Please login to your online account to display your discounted pricing

4-Fluoro-5-nitrobenzene-1,2-diamine, Technical Grade, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  BTB08542DA

 View more versions of this product

Catalog No. BTB08542DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Formula Weight 171.13
Quantity 1g
Assay Tech.
Chemical Name or Material 4-Fluoro-5-nitrobenzene-1, 2-diamine
Molecular Formula C6H6FN3O2
CAS 113269-06-0
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 4-Fluoro-5-nitrobenzene-1, 2-diamine
Assay Tech.
Molecular Formula C6H6FN3O2
Formula Weight 171.13