missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-4'-methoxybenzophenone, 98%

Manufacturer:  Alfa Aesar™ H6167903

 View more versions of this product

Catalog No. AAH6167903



Specifications

Specifications

4-Fluoro-4'-methoxybenzophenone
1g
C14H11FO2
4-fluoro-4'-methoxybenzophenone, 4-fluorophenyl 4-methoxyphenyl methanone, 4-fluorophenyl-4-methoxyphenyl methanone, 4-fluorphenyl 4-methoxyphenyl methanon, pubchem3309, acmc-209i8i, 4-methoxy-4'-fluorobenzophenone, 4-fluoro-4-methoxybenzophenone, 4-fluoro-4'-methoxy-benzophenone, 4-fluorophenyl 4-methoxyphenyl ketone
COC1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)F
230.238
230.23
98%
White
345-89-1
MFCD00055469
VWGWRNBIAWTWIB-UHFFFAOYSA-N
(4-fluorophenyl)-(4-methoxyphenyl)methanone
67664
Crystalline Powder

Chemical Identifiers

345-89-1
230.238
VWGWRNBIAWTWIB-UHFFFAOYSA-N
67664
COC1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)F
C14H11FO2
MFCD00055469
4-fluoro-4'-methoxybenzophenone, 4-fluorophenyl 4-methoxyphenyl methanone, 4-fluorophenyl-4-methoxyphenyl methanone, 4-fluorphenyl 4-methoxyphenyl methanon, pubchem3309, acmc-209i8i, 4-methoxy-4'-fluorobenzophenone, 4-fluoro-4-methoxybenzophenone, 4-fluoro-4'-methoxy-benzophenone, 4-fluorophenyl 4-methoxyphenyl ketone
(4-fluorophenyl)-(4-methoxyphenyl)methanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents