missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluoro-4'-methoxybenzophenone, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H6167914

 View more versions of this product

Catalog No. AAH6167914



Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4-Fluoro-4'-methoxybenzophenone
White
MFCD00055469
4-fluoro-4'-methoxybenzophenone, 4-fluorophenyl 4-methoxyphenyl methanone, 4-fluorophenyl-4-methoxyphenyl methanone, 4-fluorphenyl 4-methoxyphenyl methanon, pubchem3309, acmc-209i8i, 4-methoxy-4'-fluorobenzophenone, 4-fluoro-4-methoxybenzophenone, 4-fluoro-4'-methoxy-benzophenone, 4-fluorophenyl 4-methoxyphenyl ketone
COC1=CC=C(C=C1)C(=O)C1=CC=C(F)C=C1
230.24
230.23
Crystalline Powder
345-89-1
C14H11FO2
25 g
VWGWRNBIAWTWIB-UHFFFAOYSA-N
(4-fluorophenyl)-(4-methoxyphenyl)methanone
67664
98%

Chemical Identifiers

345-89-1
230.24
VWGWRNBIAWTWIB-UHFFFAOYSA-N
67664
COC1=CC=C(C=C1)C(=O)C1=CC=C(F)C=C1
C14H11FO2
MFCD00055469
4-fluoro-4'-methoxybenzophenone, 4-fluorophenyl 4-methoxyphenyl methanone, 4-fluorophenyl-4-methoxyphenyl methanone, 4-fluorphenyl 4-methoxyphenyl methanon, pubchem3309, acmc-209i8i, 4-methoxy-4'-fluorobenzophenone, 4-fluoro-4-methoxybenzophenone, 4-fluoro-4'-methoxy-benzophenone, 4-fluorophenyl 4-methoxyphenyl ketone
(4-fluorophenyl)-(4-methoxyphenyl)methanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 206-464-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions