missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-4'-methoxybenzophenone, 98%

Manufacturer:  Alfa Aesar™ H6167906

 View more versions of this product

Catalog No. AAH6167906



Specifications

Specifications

4-Fluoro-4'-methoxybenzophenone
5g
C14H11FO2
4-fluoro-4'-methoxy-benzophenone
COC1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)F
230.238
230.23
98%
White
345-89-1
MFCD00055469
VWGWRNBIAWTWIB-UHFFFAOYSA-N
(4-fluorophenyl)-(4-methoxyphenyl)methanone
67664
Crystalline Powder

Chemical Identifiers

345-89-1
230.238
VWGWRNBIAWTWIB-UHFFFAOYSA-N
67664
COC1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)F
C14H11FO2
MFCD00055469
4-fluoro-4'-methoxy-benzophenone
(4-fluorophenyl)-(4-methoxyphenyl)methanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 206-464-2

TSCA : No

Recommended Storage : Ambient temperatures

Documents