Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Fluoro-3'-(trifluoromethyl)propiophenone, 98%

$105.03 - $432.45

Chemical Identifiers

Chemical Name or Material 4'-Fluoro-3'-(trifluoromethyl)propiophenone
CAS 239107-27-8
Molecular Formula C10H8F4O
Formula Weight 220.17
MDL Number MFCD00061264
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2068503

  SDS
alfa aesar™
B2068503
1g Each for $105.03

AAB2068506

  SDS
alfa aesar™
B2068506
5g Each for $432.45
Description & Specifications

Specifications

Formula Weight 220.17
Chemical Name or Material 4'-Fluoro-3'-(trifluoromethyl)propiophenone
Assay 98%
Molecular Formula C10H8F4O
MDL Number MFCD00061264
CAS 239107-27-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 30° to 33°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.