Learn More
4-Fluoro-3-(trifluoromethyl)benzylamine, 94%, Thermo Scientific™
$107.22 - $107.22
Chemical Identifiers
CAS | 67515-74-6 |
---|---|
Molecular Formula | C8H8F4N |
Molecular Weight (g/mol) | 194.15 |
MDL Number | MFCD00070814 |
InChI Key | HZDVQEUISWBXPV-UHFFFAOYSA-O |
Synonym | 4-fluoro-3-trifluoromethyl benzylamine, 4-fluoro-3-trifluoromethyl phenyl methanamine, 4-fluoro-3-trifluoromethyl benzyl amine, 4-fluoro-3-trifluoromethylbenzylamine, 4-fluoro-3-trifluoromethyl-benzylamine, benzenemethanamine, 4-fluoro-3-trifluoromethyl, 4-fluoro-3-trifluoromethyl phenyl methylamine, 1-4-fluoro-3-trifluoromethyl phenyl methanamine, rarechem al bw 0670 |
PubChem CID | 522270 |
IUPAC Name | [4-fluoro-3-(trifluoromethyl)phenyl]methanamine |
SMILES | [NH3+]CC1=CC=C(F)C(=C1)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1024403
|
Thermo Scientific Chemicals
L1024403 |
1 g |
Each for $107.22
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
67515-74-6 | |
194.15 | |
HZDVQEUISWBXPV-UHFFFAOYSA-O | |
522270 | |
[NH3+]CC1=CC=C(F)C(=C1)C(F)(F)F |
C8H8F4N | |
MFCD00070814 | |
4-fluoro-3-trifluoromethyl benzylamine, 4-fluoro-3-trifluoromethyl phenyl methanamine, 4-fluoro-3-trifluoromethyl benzyl amine, 4-fluoro-3-trifluoromethylbenzylamine, 4-fluoro-3-trifluoromethyl-benzylamine, benzenemethanamine, 4-fluoro-3-trifluoromethyl, 4-fluoro-3-trifluoromethyl phenyl methylamine, 1-4-fluoro-3-trifluoromethyl phenyl methanamine, rarechem al bw 0670 | |
[4-fluoro-3-(trifluoromethyl)phenyl]methanamine |
Specifications
67515-74-6 | |
C8H8F4N | |
MFCD00070814 | |
UN2735 | |
Air Sensitive | |
HZDVQEUISWBXPV-UHFFFAOYSA-O | |
[4-fluoro-3-(trifluoromethyl)phenyl]methanamine | |
522270 | |
94% |
101°C (3 mmHg) | |
1.4535 | |
1 g | |
3083164 | |
4-fluoro-3-trifluoromethyl benzylamine, 4-fluoro-3-trifluoromethyl phenyl methanamine, 4-fluoro-3-trifluoromethyl benzyl amine, 4-fluoro-3-trifluoromethylbenzylamine, 4-fluoro-3-trifluoromethyl-benzylamine, benzenemethanamine, 4-fluoro-3-trifluoromethyl, 4-fluoro-3-trifluoromethyl phenyl methylamine, 1-4-fluoro-3-trifluoromethyl phenyl methanamine, rarechem al bw 0670 | |
[NH3+]CC1=CC=C(F)C(=C1)C(F)(F)F | |
194.15 | |
193.15 | |
4-Fluoro-3-(trifluoromethyl)benzylamine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon