missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-(trifluoromethyl)benzylamine, 94%

Manufacturer:  Alfa Aesar™ L1024403

 View more versions of this product

Catalog No. AAL1024403



Specifications

Specifications

4-Fluoro-3-(trifluoromethyl)benzylamine
1g
C8H7F4N
UN2735
4-fluoro-3-trifluoromethyl benzylamine, 4-fluoro-3-trifluoromethyl phenyl methanamine, 4-fluoro-3-trifluoromethyl phenyl methanamine, 4-fluoro-3-trifluoromethyl benzyl amine, 4-fluoro-3-trifluoromethylbenzylamine, 4-fluoro-3-trifluoromethyl-benzylamine, benzenemethanamine, 4-fluoro-3-trifluoromethyl, 4-fluoro-3-trifluoromethyl phenyl methylamine, 1-4-fluoro-3-trifluoromethyl phenyl methanamine, rarechem al bw 0670
C1=CC(=C(C=C1CN)C(F)(F)F)F
193.145
193.15
101°C (3mmHg)
67515-74-6
MFCD00070814
3083164
HZDVQEUISWBXPV-UHFFFAOYSA-N
[4-fluoro-3-(trifluoromethyl)phenyl]methanamine
522270
94%

Chemical Identifiers

67515-74-6
193.145
HZDVQEUISWBXPV-UHFFFAOYSA-N
522270
C1=CC(=C(C=C1CN)C(F)(F)F)F
C8H7F4N
MFCD00070814
4-fluoro-3-trifluoromethyl benzylamine, 4-fluoro-3-trifluoromethyl phenyl methanamine, 4-fluoro-3-trifluoromethyl phenyl methanamine, 4-fluoro-3-trifluoromethyl benzyl amine, 4-fluoro-3-trifluoromethylbenzylamine, 4-fluoro-3-trifluoromethyl-benzylamine, benzenemethanamine, 4-fluoro-3-trifluoromethyl, 4-fluoro-3-trifluoromethyl phenyl methylamine, 1-4-fluoro-3-trifluoromethyl phenyl methanamine, rarechem al bw 0670
[4-fluoro-3-(trifluoromethyl)phenyl]methanamine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

Documents