Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-(trifluoromethyl)benzyl alcohol, 97%

$292.46 - $292.46

Chemical Identifiers

Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)benzyl alcohol
CAS 67515-61-1
Molecular Formula C8H6F4O
Formula Weight 194.13
MDL Number MFCD00061171
Beilstein 1960409
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2081406

  SDS
alfa aesar™
B2081406
5g Each for $292.46
Description & Specifications

Specifications

Formula Weight 194.13
Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)benzyl alcohol
Assay 97%
Molecular Formula C8H6F4O
MDL Number MFCD00061171
CAS 67515-61-1
Beilstein 1960409
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 80° to 85°C (0.1mmHg)
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.