Please login to your online account to display your discounted pricing

4-Fluoro-3-(trifluoromethyl)benzoyl chloride, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC380560010

 View more versions of this product

Catalog No. AC380560010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 226.56
Quantity 1g
Color Clear colorless to pale yellow
Density 1.4930g/mL
CAS Min % 96.0
CAS Max % 100.0
Refractive Index 1.4680 to 1.4700 (20°C, 589nm)
Linear Formula FC6H3(CF3)COCl
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD00061159
Assay Percent Range 97%
Molecular Formula C8 H3 Cl F4 O
Packaging Glass bottle
Infrared Spectrum Authentic
Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)benzoyl chloride
CAS 67515-56-4
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

Specifications

Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)benzoyl chloride
CAS 67515-56-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8 H3 Cl F4 O
Linear Formula FC6H3(CF3)COCl
Formula Weight 226.56
MDL Number MFCD00061159
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

SDS & Safety Documents