Please login to your online account to display your discounted pricing

4-Fluoro-3-(trifluoromethyl)benzoyl chloride, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)benzoyl chloride
CAS 67515-56-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8 H3 Cl F4 O
Linear Formula FC6H3(CF3)COCl
Formula Weight 226.56
MDL Number MFCD00061159
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC380560010

  SDS
thermo fisher scientific
AC380560010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 226.56
Quantity 1g
Color Clear colorless to pale yellow
Density 1.4930g/mL
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula FC6H3(CF3)COCl
MDL Number MFCD00061159
Health Hazard 1 GHS Signal Word: Danger
Refractive Index 1.4680 to 1.4700 (20°C, 589nm)
Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)benzoyl chloride
Infrared Spectrum Authentic
Molecular Formula C8 H3 Cl F4 O
CAS 67515-56-4
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.