Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-(trifluoromethyl)benzoyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)benzoyl chloride
CAS 67515-56-4
Molecular Formula C8H3ClF4O
Formula Weight 226.56
MDL Number MFCD00061159
UN Number UN3094
Beilstein 1968721
Synonym alpha,alpha,alpha,4-Tetrafluoro-m-toluoyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2001603

  SDS
alfa aesar™
B2001603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2001606

  SDS
alfa aesar™
B2001606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 226.56
Flash Point 76°C (168°F)
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
MDL Number MFCD00061159
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Boiling Point 169° to 171°C
Synonym alpha,alpha,alpha,4-Tetrafluoro-m-toluoyl chloride
Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)benzoyl chloride
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
UN Number UN3094
Quantity 1g
Molecular Formula C8H3ClF4O
Assay 97%
Beilstein 1968721
Health Hazard 1 Danger
CAS 67515-56-4
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.