Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-(trifluoromethyl)benzophenone, 97+%

$67.05 - $67.05

Chemical Identifiers

Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)benzophenone
CAS 239087-04-8
Molecular Formula C14H8F4O
Formula Weight 268.21
MDL Number MFCD00061258
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2459903

  SDS
alfa aesar™
B2459903
1g Each for $67.05
Description & Specifications

Specifications

Formula Weight 268.21
Quantity 1g
Melting Point 44° to 47°C
Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)benzophenone
Molecular Formula C14H8F4O
MDL Number MFCD00061258
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 239087-04-8
Assay ≥97%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.