Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-(trifluoromethyl)benzamide, 97%

$79.24 - $79.24

Chemical Identifiers

Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)benzamide
CAS 67515-57-5
Molecular Formula C8H5F4NO
Formula Weight 207.13
MDL Number MFCD00061269
Beilstein 1968715
Synonym alpha,alpha,alpha,4-Tetrafluoro-m-toluamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2050203

  SDS
alfa aesar™
B2050203
1g Each for $79.24
Description & Specifications

Specifications

Formula Weight 207.13
Beilstein 1968715
Quantity 1g
CAS 67515-57-5
Melting Point 84° to 85°C
Assay 97%
Health Hazard 1 Warning
MDL Number MFCD00061269
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)benzamide
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C8H5F4NO
Synonym alpha,alpha,alpha,4-Tetrafluoro-m-toluamide
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.