Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-(trifluoromethyl)benzaldehyde, 97%

$943.23 - $943.23

Chemical Identifiers

Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)benzaldehyde
CAS 67515-60-0
Molecular Formula C8H4F4O
Formula Weight 192.11
MDL Number MFCD00061314
Beilstein 1958528
Synonym alpha,alpha,alpha,4-Tetrafluoro-m-tolualdehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2084414

  SDS
alfa aesar™
B2084414
25g Each for $943.23
Description & Specifications

Specifications

Formula Weight 192.11
Flash Point 76°C (168°F)
Beilstein 1958528
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 178°C
MDL Number MFCD00061314
Health Hazard 1 Warning
Quantity 25g
Synonym alpha,alpha,alpha,4-Tetrafluoro-m-tolualdehyde
Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)benzaldehyde
CAS 67515-60-0
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Assay 97%
Molecular Formula C8H4F4O
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.