Learn More
4-Fluoro-3-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals B2084414
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Fluoro-3-(trifluoromethyl)benzaldehyde | |
1.408 | |
76°C (168°F) | |
1.457 | |
25 g | |
Air Sensitive | |
BIUDHHGROGJSHN-UHFFFAOYSA-N | |
4-fluoro-3-(trifluoromethyl)benzaldehyde | |
522269 | |
97% |
67515-60-0 | |
178°C | |
C8H4F4O | |
MFCD00061314 | |
1958528 | |
4-fluoro-3-trifluoromethyl benzaldehyde, 3-trifluoromethyl-4-fluorobenzaldehyde, benzaldehyde, 4-fluoro-3-trifluoromethyl, alpha,alpha,alpha,4-tetrafluoro-m-tolualdehyde, pubchem1462, acmc-209nzm, ksc359a9l, 4-fluoro-3-trifluoromethyl | |
C1=CC(=C(C=C1C=O)C(F)(F)F)F | |
192.113 | |
192.11 |
Chemical Identifiers
67515-60-0 | |
192.113 | |
BIUDHHGROGJSHN-UHFFFAOYSA-N | |
522269 | |
C1=CC(=C(C=C1C=O)C(F)(F)F)F |
C8H4F4O | |
MFCD00061314 | |
4-fluoro-3-trifluoromethyl benzaldehyde, 3-trifluoromethyl-4-fluorobenzaldehyde, benzaldehyde, 4-fluoro-3-trifluoromethyl, alpha,alpha,alpha,4-tetrafluoro-m-tolualdehyde, pubchem1462, acmc-209nzm, ksc359a9l, 4-fluoro-3-trifluoromethyl | |
4-fluoro-3-(trifluoromethyl)benzaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures