Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-(trifluoromethyl)anisole, 97%

$62.68 - $219.99

Chemical Identifiers

Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)anisole
CAS 127271-65-2
Molecular Formula C8H6F4O
Formula Weight 194.13
MDL Number MFCD01631405
Synonym 2-Fluoro-5-methoxybenzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2456303

  SDS
alfa aesar™
B2456303
1g Each for $62.68

AAB2456306

  SDS
alfa aesar™
B2456306
5g Each for $219.99
Description & Specifications

Specifications

Formula Weight 194.13
Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)anisole
Assay 97%
Molecular Formula C8H6F4O
MDL Number MFCD01631405
CAS 127271-65-2
Synonym 2-Fluoro-5-methoxybenzotrifluoride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 67°C (153°F)
Quantity 1g

Applications
4-Fluoro-3-(trifluoromethyl)anisole is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.