Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-(trifluoromethyl)aniline, 98%

Chemical Identifiers

Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)aniline
CAS 2357-47-3
Molecular Formula C7H5F4N
Formula Weight 179.12
MDL Number MFCD00007834
UN Number UN2810
Beilstein 641587
Synonym 4-Fluoro-3-(trifluoromethyl)aniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1825006

  SDS
alfa aesar™
A1825006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1825014

  SDS
alfa aesar™
A1825014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 179.12
UN Number UN2810
Beilstein 641587
Synonym 4-Fluoro-3-(trifluoromethyl)aniline
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
MDL Number MFCD00007834
CAS 2357-47-3
Flash Point 91°C (195°F)
Molecular Formula C7H5F4N
Health Hazard 2 GHS H Statement
H311-H227-H302-H332-H315-H319
Toxic in contact with skin.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Quantity 5g
Assay 98%
Odor Characteristic
Boiling Point 74° to 75°C (10mmHg)
Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)aniline
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H227-H302-H332-H315-H319
Toxic in contact with skin.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.