Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Fluoro-3'-(trifluoromethyl)acetophenone, 97%

$62.53 - $232.04

Chemical Identifiers

Chemical Name or Material 4'-Fluoro-3'-(trifluoromethyl)acetophenone
CAS 208173-24-4
Molecular Formula C9H6F4O
Formula Weight 206.14
MDL Number MFCD00061252
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1650103

 
alfa aesar™
A1650103
1g Each for $62.53

AAA1650106

 
alfa aesar™
A1650106
5g Each for $232.04
Description & Specifications

Specifications

Formula Weight 206.14
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Chemical Name or Material 4'-Fluoro-3'-(trifluoromethyl)acetophenone
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
MDL Number MFCD00061252
CAS 208173-24-4
Molecular Formula C9H6F4O
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.