Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-(trifluoromethoxy)phenol, 97%

$148.85 - $381.74

Chemical Identifiers

Chemical Name or Material 4-Fluoro-3-(trifluoromethoxy)phenol
CAS 886501-26-4
Molecular Formula C7H4F4O2
Formula Weight 196.1
MDL Number MFCD06660257
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2676603

  SDS
alfa aesar™
H2676603
1g Each for $381.74

AAH26766MD

  SDS
alfa aesar™
H26766MD
250mg Each for $148.85
Description & Specifications

Specifications

Formula Weight 196.1
Assay 97%
Molecular Formula C7H4F4O2
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Chemical Name or Material 4-Fluoro-3-(trifluoromethoxy)phenol
MDL Number MFCD06660257
CAS 886501-26-4
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.