Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-(trifluoromethoxy)benzoic acid, 97%

$190.86 - $190.86

Chemical Identifiers

Chemical Name or Material 4-Fluoro-3-(trifluoromethoxy)benzoic acid
CAS 886496-49-7
Molecular Formula C8H4F4O3
Formula Weight 224.11
MDL Number MFCD06660161
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2636803

  SDS
alfa aesar™
H2636803
1g Each for $190.86
Description & Specifications

Specifications

Formula Weight 224.11
Chemical Name or Material 4-Fluoro-3-(trifluoromethoxy)benzoic acid
Assay 97%
Molecular Formula C8H4F4O3
MDL Number MFCD06660161
CAS 886496-49-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 86° to 90°C
Quantity 1g

Applications
4-Fluoro-3-(trifluoromethoxy)benzoic acid is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.