Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-(trifluoromethoxy)benzaldehyde, 97%

$315.55 - $954.35

Chemical Identifiers

Chemical Name or Material 4-Fluoro-3-(trifluoromethoxy)benzaldehyde
CAS 86256-48-6
Molecular Formula C8H4F4O2
Formula Weight 208.11
MDL Number MFCD06660159
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2625603

  SDS
alfa aesar™
H2625603
1g Each for $315.55

AAH2625606

  SDS
alfa aesar™
H2625606
5g Each for $954.35
Description & Specifications

Specifications

Formula Weight 208.11
Chemical Name or Material 4-Fluoro-3-(trifluoromethoxy)benzaldehyde
Assay 97%
Molecular Formula C8H4F4O2
MDL Number MFCD06660159
CAS 86256-48-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g

Applications
Used as pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is sensitive to air.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.