Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-(trifluoromethoxy)aniline, 97%

Chemical Identifiers

Chemical Name or Material 4-Fluoro-3-(trifluoromethoxy)aniline
Molecular Formula C7H5F4NO
Formula Weight 195.12
MDL Number MFCD09832246
UN Number UN2810
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3281003

  SDS
alfa aesar™
H3281003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH32810MD

  SDS
alfa aesar™
H32810MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 195.12
UN Number UN2810
Molecular Formula C7H5F4NO
MDL Number MFCD09832246
Assay 97%
Health Hazard 1 Danger
Quantity 1g
Health Hazard 2 GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Chemical Name or Material 4-Fluoro-3-(trifluoromethoxy)aniline

Applications
4-Fluoro-3-(trifluoromethyl)aniline is used in the synthesis of trifluoromethylated poly(phenylene oxide). It is used in the preparation of 2-[3-(4-fluoro-3-trifluoromethyl-phenyl)-ureido]-2-methyl-propionic acid methyl ester by reacting with 2-isocyanato-2-methyl-propionic acid methyl ester. It is also used as a chemical intermediate in the synthesis of agrochemicals, pharmaceuticals and dyes.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and acids.

Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.