Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Fluoro-3'-(trifluoromethoxy)acetophenone, 97%

$200.11 - $601.86

Chemical Identifiers

Chemical Name or Material 4'-Fluoro-3'-(trifluoromethoxy)acetophenone
CAS 886501-44-6
Molecular Formula C9H6F4O2
Formula Weight 222.14
MDL Number MFCD06660260
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2684403

 
alfa aesar™
H2684403
1g Each for $200.11

AAH2684406

 
alfa aesar™
H2684406
5g Each for $601.86
Description & Specifications

Specifications

Formula Weight 222.14
Chemical Name or Material 4'-Fluoro-3'-(trifluoromethoxy)acetophenone
Assay 97%
Molecular Formula C9H6F4O2
MDL Number MFCD06660260
CAS 886501-44-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.