missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Fluoro-3-(tetrazol-5-yl)phenylboronic acid, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 445460010
Specifications
4-Fluoro-3-(tetrazol-5-yl)phenylboronic acid | |
94% min. (HPLC) | |
C7H6BFN4O2 | |
1g | |
FELFJHZMQYCKPT-UHFFFAOYSA-N | |
[4-fluoro-3-(2H-tetrazol-5-yl)phenyl]boronic acid | |
11287236 | |
95% |
1009303-56-3 | |
Glass Bottle | |
MFCD13176533 | |
4-fluoro-3-tetrazol-5-yl phenylboronic acid, 4-fluoro-3-1h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 4-fluoro-3-1h-tetrazol-5-yl phenyl boronic acid, 4-fluoro-3-1h-tetrazol-5-yl phenylboronic acid | |
OB(O)C1=CC(C2=NNN=N2)=C(F)C=C1 | |
207.96 | |
207.96 |
Chemical Identifiers
1009303-56-3 | |
207.96 | |
FELFJHZMQYCKPT-UHFFFAOYSA-N | |
11287236 | |
OB(O)C1=CC(C2=NNN=N2)=C(F)C=C1 |
C7H6BFN4O2 | |
MFCD13176533 | |
4-fluoro-3-tetrazol-5-yl phenylboronic acid, 4-fluoro-3-1h-1,2,3,4-tetrazol-5-yl phenylboronic acid, 4-fluoro-3-1h-tetrazol-5-yl phenyl boronic acid, 4-fluoro-3-1h-tetrazol-5-yl phenylboronic acid | |
[4-fluoro-3-(2H-tetrazol-5-yl)phenyl]boronic acid |
Safety and Handling
GHS Signal Word: Warning
RUO â Research Use Only