Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-(phenoxycarbonylamino)benzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 4-Fluoro-3-(phenoxycarbonylamino)benzeneboronic acid pinacol ester
Formula Weight 357.1
Synonym 4-Fluoro-3-(phenoxycarbonylamino)phenylboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6283703

 
alfa aesar™
H6283703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62837MD

 
alfa aesar™
H62837MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 357.1
Physical Form Solid
Synonym 4-Fluoro-3-(phenoxycarbonylamino)phenylboronic acid pinacol ester
Quantity 1g
Chemical Name or Material 4-Fluoro-3-(phenoxycarbonylamino)benzeneboronic acid pinacol ester
Assay 96%