Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Fluoro-3-phenoxybenzaldehyde, 97%

Chemical Identifiers

Chemical Name or Material 4-Fluoro-3-phenoxybenzaldehyde
CAS 68359-57-9
Formula Weight 216.21
MDL Number MFCD01318148
UN Number UN3082
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6120206

  SDS
alfa aesar™
H6120206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6120214

  SDS
alfa aesar™
H6120214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 216.21
Chemical Name or Material 4-Fluoro-3-phenoxybenzaldehyde
TSCA TSCA
Physical Form Liquid
Assay 97%
MDL Number MFCD01318148
CAS 68359-57-9
UN Number UN3082
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Refractive Index 1.583
DOT Information Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
Density 1.209g/mL
Color Colorless
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.