missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluoro-3-nitrobenzotrifluoride, TCI America™

Manufacturer:  TCI America F0324250G

 View more versions of this product

Catalog No. F0324250G



Specifications

Specifications

4-Fluoro-3-nitrobenzotrifluoride
Undesignated
367-86-2
MFCD00007059
4-fluoro-3-nitrobenzotrifluoride, 1-fluoro-2-nitro-4-trifluoromethyl benzene, benzene, 1-fluoro-2-nitro-4-trifluoromethyl, 4-fluoro-3-nitrotrifluoromethylbenzene, 3-nitro-4-fluorobenzotrifluoride, 3-nitro-alpha,alpha,alpha,4-tetrafluorotoluene, 2-fluoro-5-trifluoromethyl nitrobenzene, 3-nitro-4-fluoro benzotrifluoride, 2-fluoro-5-trifluoromethyl nitrobenzene, 1-fluoro-2-nitro-4-trifluoromethyl-benzene
C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])F
209.1
209.10
≥95.0% (GC)
208°C
250g
C7H3F4NO2
1993
HLDFCCHSOZWKAA-UHFFFAOYSA-N
1-fluoro-2-nitro-4-(trifluoromethyl)benzene
67778
Clear Liquid at 20°C

Chemical Identifiers

367-86-2
209.1
HLDFCCHSOZWKAA-UHFFFAOYSA-N
67778
C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])F
C7H3F4NO2
MFCD00007059
4-fluoro-3-nitrobenzotrifluoride, 1-fluoro-2-nitro-4-trifluoromethyl benzene, benzene, 1-fluoro-2-nitro-4-trifluoromethyl, 4-fluoro-3-nitrotrifluoromethylbenzene, 3-nitro-4-fluorobenzotrifluoride, 3-nitro-alpha,alpha,alpha,4-tetrafluorotoluene, 2-fluoro-5-trifluoromethyl nitrobenzene, 3-nitro-4-fluoro benzotrifluoride, 2-fluoro-5-trifluoromethyl nitrobenzene, 1-fluoro-2-nitro-4-trifluoromethyl-benzene
1-fluoro-2-nitro-4-(trifluoromethyl)benzene
Safety and Handling

Safety and Handling

TSCA : No

Documents